CAS RN 1215566-78-1
IUPAC Name:
6-chloro-5-methyl-N-{6-[(2-methylpyridin-3-yl)oxy]pyridin-3-yl}-2,3-dihydro-1H-indole-1-carboxamide dihydrochloride
Synonyms:
6-chloro-5-methyl-N-{6-[(2-methylpyridin-3-yl)oxy]pyridin-3-yl}-2,3-dihydroindole-1-carboxamide dihydrochloride
Molecular Weight (g/mol):
467.78
Molecular Formula:
C21H21Cl3N4O2
InChi Key:
GCMNSEILNIPNSX-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC1=C(Cl)C=C2N(CCC2=C1)C(=O)NC1=CN=C(OC2=C(C)N=CC=C2)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ SB 242084
CAS: 1215566-78-1 Molecular Formula: C21H21Cl3N4O2