Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 242084
GREENER_CHOICESDP

Catalog No. 290150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-015-0 50 mg
29-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-015-0 Supplier Tocris Bioscience™ Supplier No. 2901/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective 5-HT2C antagonist; brain penetrant

5-HT2C receptor antagonist that displays 158- and 100-fold selectivity over 5-HT2A and 5-HT2B receptors respectively. Also displays selectivity over a range of other 5-HT, dopamine and adrenergic receptors. Brain penetrant; exerts anxiolytic-like activity.

Chemical Identifiers

CAS 1215566-78-1
Molecular Formula C21H21Cl3N4O2

Specifications

Quantity 50 mg
Formula Weight 467.78
Purity >99%
Molecular Weight (g/mol) 467.775
Chemical Name or Material SB 242084
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.