CAS RN 1217756-94-9
IUPAC Name:
N-[3-(dimethylamino)-2,3,4,9-tetrahydro-1H-carbazol-6-yl]-4-fluorobenzamide hydrochloride
Synonyms:
N-[6-(dimethylamino)-6,7,8,9-tetrahydro-5H-carbazol-3-yl]-4-fluorobenzamide hydrochloride
Molecular Weight (g/mol):
387.88
Molecular Formula:
C21H23ClFN3O
InChi Key:
OKUHLSYESBLBCP-UHFFFAOYNA-N
SMILES:
Cl.CN(C)C1CCC2=C(C1)C1=CC(NC(=O)C3=CC=C(F)C=C3)=CC=C1N2
1
–
1
of
1
results
1
R&D Systems™ LY 344864 hydrochloride
CAS: 1217756-94-9 Molecular Formula: C21H23ClFN3O