CAS RN 1219810-16-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ 1S,3R-RSL3
Induces ferroptosis; inhibits glutathione peroxidase 4 (GPX4)