CAS RN 1222781-70-5
IUPAC Name:
[(4-methoxyphenyl)methyl][(naphthalen-1-yl)methyl]amine hydrochloride
Synonyms:
[(4-methoxyphenyl)methyl](naphthalen-1-ylmethyl)amine hydrochloride
Molecular Weight (g/mol):
313.83
Molecular Formula:
C19H20ClNO
InChi Key:
NGQIBUUFXDPHKT-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=C(CNCC2=C3C=CC=CC3=CC=C2)C=C1
1
–
1
of
1
results
1
R&D Systems™ ML 133 hydrochloride
CAS: 1222781-70-5 Molecular Formula: C19H20ClNO