accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNNONNFFF

CAS RN 1222998-36-8

IUPAC Name: 1-[4-(4-propanoylpiperazin-1-yl)-3-(trifluoromethyl)phenyl]-9-(quinolin-3-yl)-1H,2H-benzo[h]1,6-naphthyridin-2-one
Synonyms: 1-[4-(4-propanoylpiperazin-1-yl)-3-(trifluoromethyl)phenyl]-9-(quinolin-3-yl)benzo[h]1,6-naphthyridin-2-one
Molecular Weight (g/mol): 607.64
Molecular Formula: C35H28F3N5O2
InChi Key: AKCRNFFTGXBONI-UHFFFAOYSA-N
SMILES: CCC(=O)N1CCN(CC1)C1=C(C=C(C=C1)N1C(=O)C=CC2=C1C1=CC(=CC=C1N=C2)C1=CN=C2C=CC=CC2=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results