accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONNNH2FFF

CAS RN 1223001-51-1

IUPAC Name: 9-(6-aminopyridin-3-yl)-1-[3-(trifluoromethyl)phenyl]-1H,2H-benzo[h]1,6-naphthyridin-2-one
Synonyms: 9-(6-aminopyridin-3-yl)-1-[3-(trifluoromethyl)phenyl]benzo[h]1,6-naphthyridin-2-one
Molecular Weight (g/mol): 432.41
Molecular Formula: C24H15F3N4O
InChi Key: GUXXEUUYCAYESJ-UHFFFAOYSA-N
SMILES: NC1=NC=C(C=C1)C1=CC=C2N=CC3=C(N(C(=O)C=C3)C3=CC=CC(=C3)C(F)(F)F)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results