Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Torin 2
GREENER_CHOICESDP

Catalog No. 424850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-485-0 50 mg
42-481-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-485-0 Supplier R&D Systems™ Supplier No. 4248/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective mTOR inhibitor

Potent and selective mTOR inhibitor (IC50 = 2.1 nM). Displays 800-fold cellular selectivity for mTOR over PI3K (cellular EC50 values are 0.25 and 200 nM for mTOR and PI3K respectively). Activates autophagy. Orally available.

Chemical Identifiers

CAS 1223001-51-1
Molecular Formula C24H15F3N4O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 432.4
Purity >98%
Molecular Weight (g/mol) 432.406
Chemical Name or Material Torin 2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.