CAS RN 1223397-11-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Glucose Transporter Inhibitor IV, WZB117
The Glucose Transporter Inhibitor IV, WZB117 controls the biological activity of Glucose Transporter.
2
Tocris Bioscience™ WZB 117
GLUT inhibitor