accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOSNNHNN

CAS RN 1226056-71-8

IUPAC Name: N-benzyl-2-[(pyrimidin-4-yl)amino]-1,3-thiazole-4-carboxamide
Synonyms: N-benzyl-2-(pyrimidin-4-ylamino)-1,3-thiazole-4-carboxamide
Molecular Weight (g/mol): 311.36
Molecular Formula: C15H13N5OS
InChi Key: DOBKQCZBPPCLEG-UHFFFAOYSA-N
SMILES: O=C(NCC1=CC=CC=C1)C1=CSC(NC2=NC=NC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results