Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Thiazovivin
GREENER_CHOICESDP

Catalog No. 384510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 38-451-0 Supplier R&D Systems™ Supplier No. 3845/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. ROCK inhibitor ; improves the efficiency of fibroblast reprogramming and induction of iPSCs

Thiazovivin is a selective, cell-permeable Rho-associated coiled-coil containing protein kinase (ROCK) inhibitor (IC50 = 0.5 μM). Thiazovivin enhances the efficiency of fibroblast reprogramming to generate induced pluripotent stem cells (iPSCs) when used in combination with SB 431542 (Cat. No. 1614) and PD 0325901 (Cat. No. 4192). Thiazovivin improves the survival of human embryonic stem cells (hESCs) upon trypsinization and increases cell adhesion through the regulation of E-cadherin and significantly improves direct reprogramming efficiency. In combination with Valproic acid, sodium salt (Cat. No. 2815), Purmorphamine (Cat. No. 4551) and A 83-01 (Cat. No. 2939), Thiazovivin can be used to directly reprogram mouse embryonic fibroblasts into chemically induced neural stem cells.

Chemical Identifiers

CAS 1226056-71-8
Molecular Formula C15H13N5OS

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 311.36
Purity 98%
Molecular Weight (g/mol) 311.363
Chemical Name or Material Thiazovivin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.