accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHNNFFFF

CAS RN 1229096-88-1

IUPAC Name: N-{4-[1-(4-fluorophenyl)-1H-indazol-5-yl]-3-(trifluoromethyl)phenyl}-1-phenylmethanesulfonamide
Synonyms: N-{4-[1-(4-fluorophenyl)indazol-5-yl]-3-(trifluoromethyl)phenyl}-1-phenylmethanesulfonamide
Molecular Weight (g/mol): 525.52
Molecular Formula: C27H19F4N3O2S
InChi Key: DXBJGDVBQPEMOB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N1N=CC2=CC(=CC=C12)C1=C(C=C(NS(=O)(=O)CC2=CC=CC=C2)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results