CAS RN 1233533-04-4
IUPAC Name:
4-hydroxy-8,14-dithia-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Synonyms:
4-hydroxy-8,14-dithia-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one
Molecular Weight (g/mol):
251.32
Molecular Formula:
C11H9NO2S2
InChi Key:
DHUAGGSHTKPOHU-UHFFFAOYSA-N
SMILES:
OC1=CC=C2SC3=C(SCCNC3=O)C2=C1