accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123808-97-9

H3CCH3(R)O(S)CH3ON

CAS RN 123808-97-9

IUPAC Name: (3R,7aS)-7a-methyl-3-(propan-2-yl)-hexahydropyrrolo[2,1-b][1,3]oxazol-5-one
Synonyms: (3R,7aS)-3-isopropyl-7a-methyl-tetrahydropyrrolo[2,1-b][1,3]oxazol-5-one
Molecular Weight (g/mol): 183.25
Molecular Formula: C10H17NO2
InChi Key: NUUDVADIQSLTCN-WPRPVWTQSA-N
SMILES: CC(C)[C@@H]1CO[C@@]2(C)CCC(=O)N12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results