accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHPHOHOHO

CAS RN 124-64-1

IUPAC Name: tetrakis(hydroxymethyl)phosphanium chloride
Synonyms: tetrakis(hydroxymethyl)phosphonium chloride
Molecular Weight (g/mol): 190.56
Molecular Formula: C4H12ClO4P
InChi Key: AKXUUJCMWZFYMV-UHFFFAOYSA-M
SMILES: [Cl-].OC[P+](CO)(CO)CO
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
Boiling Point 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)