CAS RN 1245907-03-2
IUPAC Name:
2-[(6-hydroxy-6-methylheptan-2-yl)amino]-4-methyl-6-[(5-methyl-1H-pyrazol-3-yl)amino]pyridine-3-carbonitrile hydrochloride
Synonyms:
2-[(6-hydroxy-6-methylheptan-2-yl)amino]-4-methyl-6-[(5-methyl-1H-pyrazol-3-yl)amino]pyridine-3-carbonitrile hydrochloride
Molecular Weight (g/mol):
392.93
Molecular Formula:
C19H29ClN6O
InChi Key:
DXVATYHPJJPMIN-UHFFFAOYNA-N
SMILES:
Cl.CC(CCCC(C)(C)O)NC1=C(C#N)C(C)=CC(NC2=NNC(C)=C2)=N1
1
–
1
of
1
results
1
R&D Systems™ TC-A 2317 hydrochloride
CAS: 1245907-03-2 Molecular Formula: C19H29ClN6O