accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOOSOOH3COHCH3

CAS RN 125-31-5

IUPAC Name: 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: xylenol blue
Molecular Weight (g/mol): 410.48
Molecular Formula: C23H22O5S
InChi Key: MGUKYHHAGPFJMC-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C=C1C
Quantity 
  • (2)
  • (1)
Color 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)