CAS RN 125464-42-8
CAS RN 125464-42-8
IUPAC Name:
3-amino-2-(4-chlorophenyl)propane-1-sulfonic acid
Synonyms:
saclofen
Molecular Weight (g/mol):
249.71
Molecular Formula:
C9H12ClNO3S
InChi Key:
JYLNVJYYQQXNEK-UHFFFAOYNA-N
SMILES:
NCC(CS(O)(=O)=O)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Saclofen, Tocris Bioscience™


CAS: 125464-42-8 Molecular Formula: C9H12ClNO3S Molecular Weight (g/mol): 249.71