CAS RN 125572-93-2
IUPAC Name:
hydrogen (6S)-6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol chloride
Synonyms:
hydrogen neupro chloride
Molecular Weight (g/mol):
351.93
Molecular Formula:
C19H26ClNOS
InChi Key:
CEXBONHIOKGWNU-NTISSMGPSA-N
SMILES:
[H+].[Cl-].CCCN(CCC1=CC=CS1)[C@H]1CCC2=C(O)C=CC=C2C1
1
–
1
of
1
results
1
R&D Systems™ Rotigotine hydrochloride
CAS: 125572-93-2 Molecular Formula: C19H26ClNOS