CAS RN 1259314-65-2
IUPAC Name:
N-[4-(4-methylpiperazin-1-yl)phenyl]acridin-9-amine dihydrochloride
Synonyms:
N-[4-(4-methylpiperazin-1-yl)phenyl]acridin-9-amine dihydrochloride
Molecular Weight (g/mol):
441.4
Molecular Formula:
C24H26Cl2N4
InChi Key:
VVZOADYFJAJZGL-UHFFFAOYSA-N
SMILES:
Cl.Cl.CN1CCN(CC1)C1=CC=C(NC2=C3C=CC=CC3=NC3=CC=CC=C23)C=C1
1
–
1
of
1
results
1
R&D Systems™ JP 1302 dihydrochloride
CAS: 1259314-65-2 Molecular Formula: C24H26Cl2N4