Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ JP 1302 dihydrochloride
GREENER_CHOICESDP

Catalog No. 266650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-665-0 50 mg
26-661-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-665-0 Supplier R&D Systems™ Supplier No. 2666/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective α2C antagonist

α2C-adrenoceptor antagonist that displays ∽ 50-fold selectivity over other α2-adrenoceptor subtypes (Ki values are 28, 1470, 1700 and 3150 nM for human α2C, α2B, α2D and α2A subtypes respectively). Potently antagonizes adrenalin-stimulated 35GTPγS binding in vitro (KB = 16 nM) and produces antidepressant and antipsychotic-like effects in vivo.

Chemical Identifiers

CAS 1259314-65-2
Molecular Formula C24H26Cl2N4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 441.4
Purity >99%
Molecular Weight (g/mol) 441.4
Chemical Name or Material JP 1302 dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.