CAS RN 125941-87-9
IUPAC Name:
8-chloro-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepin-7-ol hydrochloride
Synonyms:
8-chloro-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrochloride
Molecular Weight (g/mol):
324.25
Molecular Formula:
C17H19Cl2NO
InChi Key:
OYCAEWMSOPMASE-UHFFFAOYNA-N
SMILES:
Cl.CN1CCC2=CC(Cl)=C(O)C=C2C(C1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ SCH 23390 hydrochloride
CAS: 125941-87-9 Molecular Formula: C17H19Cl2NO