CAS RN 1263283-43-7
IUPAC Name:
4-[2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione hydrochloride
Synonyms:
razoxane hydrochloride
Molecular Weight (g/mol):
304.73
Molecular Formula:
C11H17ClN4O4
InChi Key:
BIFMNMPSIYHKDN-UHFFFAOYNA-N
SMILES:
Cl.CC(CN1CC(=O)NC(=O)C1)N1CC(=O)NC(=O)C1
1
–
1
of
1
results
1
R&D Systems™ Dexrazoxane hydrochloride
CAS: 1263283-43-7 Molecular Formula: C11H17ClN4O4