accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONONHNNOCl

CAS RN 1266212-81-0

IUPAC Name: 5-(4-butoxy-3-chlorophenyl)-N-{[2-(morpholin-4-yl)pyridin-3-yl]methyl}pyridine-3-carboxamide
Synonyms: 5-(4-butoxy-3-chlorophenyl)-N-{[2-(morpholin-4-yl)pyridin-3-yl]methyl}pyridine-3-carboxamide
Molecular Weight (g/mol): 480.99
Molecular Formula: C26H29ClN4O3
InChi Key: JPJGFWKHSMUKFO-UHFFFAOYSA-N
SMILES: CCCCOC1=C(Cl)C=C(C=C1)C1=CC(=CN=C1)C(=O)NCC1=C(N=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results