Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 887826
GREENER_CHOICESDP

Catalog No. 424950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-495-0 50 mg
42-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-495-0 Supplier R&D Systems™ Supplier No. 4249/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent voltage-dependent NaV1.8 channel blocker

Potent voltage-dependent NaV1.8 channel blocker (IC50 = 11 nM in recombinant human NaV1.8 channels). Reduces tactile allodynia in neuropathic rat models. 3- and 28-fold more potent at NaV1.8 channels compared to NaV1.2 and NaV1.7 channels respectively.

Chemical Identifiers

CAS 1266212-81-0
Molecular Formula C26H29ClN4O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 480.99
Purity >97%
Molecular Weight (g/mol) 480.993
Chemical Name or Material A 887826
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.