accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOH

CAS RN 1274892-05-5

IUPAC Name: 10-(benzylamino)decan-1-ol
Synonyms: 10-(benzylamino)decan-1-ol
Molecular Weight (g/mol): 263.43
Molecular Formula: C17H29NO
InChi Key: PZKWYQBXXXKOOF-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results