accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFF

CAS RN 129322-83-4

IUPAC Name: 1-(2,4,5-trifluorophenyl)ethan-1-one
Synonyms: 1-(2,4,5-trifluorophenyl)ethanone
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: GVTLJUZWNNFHMZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(F)C=C1F
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)