CAS RN 129830-38-2
IUPAC Name:
4-(1-aminoethyl)-N-(pyridin-4-yl)cyclohexane-1-carboxamide dihydrochloride
Synonyms:
4-(1-aminoethyl)-N-(pyridin-4-yl)cyclohexane-1-carboxamide dihydrochloride
Molecular Weight (g/mol):
320.26
Molecular Formula:
C14H23Cl2N3O
InChi Key:
IDDDVXIUIXWAGJ-UHFFFAOYNA-N
SMILES:
Cl.Cl.CC(N)C1CCC(CC1)C(=O)NC1=CC=NC=C1
1
–
3
of
3
results
1
R&D Systems™ Y-27632 in solution
A Tocris product, part of R&D Systems. Sterile-filtered 10mM solution of Y-27632 pre-dissolved in water
2
R&D Systems™ Y-27632 dihydrochloride
A Tocris product, part of R&D Systems. Y-27632 synthesized to cGMP guidelines
3
R&D Systems™ Y-27632 dihydrochloride
CAS: 129830-38-2 Molecular Formula: C14H23Cl2N3O