Learn More
Description
For more information about how Y-27632 may be used, please see our protocols: Transdifferentiating Fibroblasts into Neurons, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts .
Research use only Y-27632 also available.
Specifications
Specifications
| Chemical Name or Material | trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride |
| CAS | 129830-38-2 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Rho-kinase Inhibitors |
| Molecular Formula | C14H21N3O.2HCl |
| Purity | 99% |
| Regulatory Status | GMP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
