CAS RN 130209-90-4
IUPAC Name:
methyl 2-(2-chlorophenyl)-2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}acetate hydrochloride
Synonyms:
clopidogrel hydrochloride
Molecular Weight (g/mol):
358.28
Molecular Formula:
C16H17Cl2NO2S
InChi Key:
XIHVAFJSGWDBGA-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N1CCC2=C(C1)C=CS2)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
R&D Systems™ (±)-Clopidogrel hydrochloride
CAS: 130209-90-4 Molecular Formula: C16H17Cl2NO2S