accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)NHNHOOOHN(R)HNOHOO

CAS RN 130332-27-3

IUPAC Name: 3-{[(1R)-2-[(2R)-2-{[(adamantan-2-yloxy)carbonyl]amino}-2-[(1H-indol-3-yl)methyl]propanamido]-1-phenylethyl]carbamoyl}propanoic acid
Synonyms: 3-{[(1R)-2-[(2R)-2-{[(adamantan-2-yloxy)carbonyl]amino}-2-(1H-indol-3-ylmethyl)propanamido]-1-phenylethyl]carbamoyl}propanoic acid
Molecular Weight (g/mol): 614.74
Molecular Formula: C35H42N4O6
InChi Key: FVQSSYMRZKLFDR-ZABPBAJSSA-N
SMILES: C[C@](CC1=CNC2=CC=CC=C12)(NC(=O)OC1C2CC3CC(C2)CC1C3)C(=O)NC[C@H](NC(=O)CCC(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results