Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CI 988
GREENER_CHOICESDP

Catalog No. 260750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-075-0 50 mg
26-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-075-0 Supplier Tocris Bioscience™ Supplier No. 2607/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CCK2 antagonist

Potent and selective CCK2 (CCK-B) receptor antagonist that displays ∽ 1600-fold selectivity over CCK1 receptors (IC50 values are 1.7 and 2717 nM for CCK2 and CCK1 respectively). Has negligible affinity at a range of other binding sites (IC50 > 10 μM). Exhibits anxiolytic activity following oral administration.

Chemical Identifiers

CAS 130332-27-3
Molecular Formula C35H42N4O6

Specifications

Quantity 50 mg
Formula Weight 614.73
Purity >99%
Molecular Weight (g/mol) 614.743
Chemical Name or Material CI 988
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.