accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130406-30-3

OHOONH(R)

CAS RN 130406-30-3

IUPAC Name: (9H-fluoren-9-yl)methyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 373.45
Molecular Formula: C24H23NO3
InChi Key: SJGBJASOHDROCR-GOSISDBHSA-N
SMILES: OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)