accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONONNOCH3

CAS RN 130477-52-0

IUPAC Name: ethyl 15-methoxy-12-oxo-2,4,11-triazatetracyclo[11.4.0.0²,⁶.0⁷,¹¹]heptadeca-1(13),3,5,14,16-pentaene-5-carboxylate
Synonyms: ethyl 15-methoxy-12-oxo-2,4,11-triazatetracyclo[11.4.0.0²,⁶.0⁷,¹¹]heptadeca-1(13),3,5,14,16-pentaene-5-carboxylate
Molecular Weight (g/mol): 341.37
Molecular Formula: C18H19N3O4
InChi Key: YKYOQIXTECBVBB-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C2C3CCCN3C(=O)C3=C(C=CC(OC)=C3)N2C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)