CAS RN 13071-11-9
IUPAC Name:
hydrogen (2R)-1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms:
D-propranolol hydrochloride dexpropranolol hcl dexpropranolol hydrochloride
Molecular Weight (g/mol):
295.81
Molecular Formula:
C16H22ClNO2
InChi Key:
ZMRUPTIKESYGQW-PFEQFJNWSA-N
SMILES:
[H+].[Cl-].CC(C)NC[C@@H](O)COC1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
R&D Systems™ (R)-(+)-Propranolol hydrochloride
CAS: 13071-11-9 Molecular Formula: C16H22ClNO2