accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COSNOH3CONNH3C

CAS RN 130717-51-0

IUPAC Name: hydrogen 1-[4,5-bis(4-methoxyphenyl)-1,3-thiazole-2-carbonyl]-4-methylpiperazine chloride
Synonyms: hydrogen 1-[4,5-bis(4-methoxyphenyl)-1,3-thiazole-2-carbonyl]-4-methylpiperazine chloride
Molecular Weight (g/mol): 459.99
Molecular Formula: C23H26ClN3O3S
InChi Key: YWMAVHIKOAOSFM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(C=C1)C1=C(N=C(S1)C(=O)N1CCN(C)CC1)C1=CC=C(OC)C=C1
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results