CAS RN 1309871-50-8
IUPAC Name:
tetrakis(triethylazanium) {[(3,4-dihydroxy-5-{2-oxo-4-[(3-phenylpropoxy)amino]-1,2-dihydropyrimidin-1-yl}oxolan-2-yl)methyl phosphonato]oxy}(phosphonatooxy)phosphinate
Synonyms:
tetrakis(TEAH) [(3,4-dihydroxy-5-{2-oxo-4-[(3-phenylpropoxy)amino]pyrimidin-1-yl}oxolan-2-yl)methyl phosphonato]oxy(phosphonatooxy)phosphinate
Molecular Weight (g/mol):
1022.11
Molecular Formula:
C42H86N7O15P3
InChi Key:
ZCDZFMKRHPANNI-UHFFFAOYNA-N
SMILES:
CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.OC1C(O)C(OC1COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)N1C=CC(NOCCCC2=CC=CC=C2)=NC1=O
1
–
1
of
1
results
1
R&D Systems™ MRS 4062 triethylammonium salt
CAS: 1309871-50-8 Molecular Formula: C42H86N7O15P3