accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHCH3NHOCHHNO(E/Z)

CAS RN 1311143-71-1

IUPAC Name: N-{3-[(2-ethyl-5-methyl-1H-imidazol-4-yl)methylidene]-2-oxo-2,3-dihydro-1H-indol-5-yl}prop-2-ynamide
Synonyms: N-{3-[(2-ethyl-5-methyl-1H-imidazol-4-yl)methylidene]-2-oxo-1H-indol-5-yl}prop-2-ynamide
Molecular Weight (g/mol): 320.35
Molecular Formula: C18H16N4O2
InChi Key: ORKOHXAJFGRZCL-UHFFFAOYSA-N
SMILES: CCC1=NC(C=C2C(=O)NC3=C2C=C(NC(=O)C#C)C=C3)=C(C)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results