CAS RN 1313613-09-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ AMZ 30
Potent, selective and irreversible protein phosphatase methylesterase-1 (PME-1) inhibitor