CAS RN 131780-48-8
IUPAC Name:
8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-oxo-1,2,3,4-tetrahydroquinazoline-3-carboxylate hydrochloride
Synonyms:
8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride
Molecular Weight (g/mol):
351.83
Molecular Formula:
C17H22ClN3O3
InChi Key:
FDERDDSQHZRNGC-UHFFFAOYNA-N
SMILES:
Cl.CN1C2CCC1CC(C2)OC(=O)N1CC2=CC=CC=C2NC1=O
1
–
1
of
1
results
1
R&D Systems™ DAU 5884 hydrochloride
CAS: 131780-48-8 Molecular Formula: C17H22ClN3O3