Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DAU 5884 hydrochloride
SDP

Catalog No. 209650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-965-0 50 mg
20-961-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-965-0 Supplier R&D Systems™ Supplier No. 2096/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. M3 receptor antagonist

Selective muscarinic M3 receptor antagonist. In bovine tracheal smooth muscle, inhibits methacholine-dependent effects on cell proliferation and muscle contractility.

Chemical Identifiers

CAS 131780-48-8
Molecular Formula C17H22ClN3O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 351.83
Purity >99%
Molecular Weight (g/mol) 351.831
Chemical Name or Material DAU 5884 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.