accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13242-55-2

CH3OOOOOCH3OOH3CO

CAS RN 13242-55-2

IUPAC Name: 2,3-bis(acetyloxy)-6,8-dioxabicyclo[3.2.1]octan-4-yl acetate
Synonyms: 2,3-bis(acetyloxy)-6,8-dioxabicyclo[3.2.1]octan-4-yl acetate
Molecular Weight (g/mol): 288.25
Molecular Formula: C12H16O8
InChi Key: BAKQMOSGYGQJOJ-UHFFFAOYNA-N
SMILES: CC(=O)OC1C2COC(O2)C(OC(C)=O)C1OC(C)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results