accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN(R)(R)OH(S)OH(R)HOONOOHN(Z)

CAS RN 132489-69-1

IUPAC Name: [(2Z,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-ylidene]amino N-phenylcarbamate
Synonyms: pugnac
Molecular Weight (g/mol): 353.33
Molecular Formula: C15H19N3O7
InChi Key: PBLNJFVQMUMOJY-JXZOILRNSA-N
SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O\C1=N/OC(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results