accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13250-82-3

IUPAC Name: 2-(thiophen-3-yl)-1,3-dioxolane
Synonyms: 2-(thiophen-3-yl)-1,3-dioxolane
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: VOJKCZSSUHBMKS-UHFFFAOYSA-N
SMILES: C1COC(O1)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results