Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Thienyl)-1,3-dioxolane 97.0+%, TCI America™
SDP

Catalog No. T26235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26235G Supplier TCI America Supplier No. T26235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13250-82-3
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD06209359
InChI Key VOJKCZSSUHBMKS-UHFFFAOYSA-N
Synonym 3-(1,3-Dioxolan-2-yl)thiophene
PubChem CID 315630
IUPAC Name 2-(thiophen-3-yl)-1,3-dioxolane
SMILES C1COC(O1)C1=CSC=C1

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 156.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Thienyl)-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.