accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3SNHN

CAS RN 132539-06-1

IUPAC Name: 5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
Synonyms: symbyax zyprexa intramuscular olansek zyprexa zydis zydis
Molecular Weight (g/mol): 312.44
Molecular Formula: C17H20N4S
InChi Key: WXPNDRBBWZMPQG-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)