Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Olanzapine
GREENER_CHOICESDP

Catalog No. 434950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-495-0 50 mg
43-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-495-0 Supplier R&D Systems™ Supplier No. 4349/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-HT2A/D2 antagonist; atypical antipsychotic

Antagonist of 5-HT2A and dopamine D2 receptors (Ki values are 4 and 11 nM respectively). Also displays affinity for a range of other receptors including D1 and D4, 5-HT2C, α1, H1 and M1-4 receptors (Ki values range between 1.9 and 31 nM) . Atypical antipsychotic. Displays anticholinergic properties. Also a highly potent activator of hM4Di DREADDs (EC50 values are 5 nM in vitro and 7 nM in vivo).

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17H20N4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 312.43
Purity >99%
Molecular Weight (g/mol) 312.44
Chemical Name or Material Olanzapine
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.