accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NNH3CNCH3NCH3(E/Z)

CAS RN 133059-99-1

IUPAC Name: hydrogen N-ethyl-1,2-dimethyl-6-(methylimino)-N-phenyl-1,6-dihydropyrimidin-4-amine chloride
Synonyms: hydrogen N-ethyl-1,2-dimethyl-6-(methylimino)-N-phenylpyrimidin-4-amine chloride
Molecular Weight (g/mol): 292.81
Molecular Formula: C15H21ClN4
InChi Key: DUWKUHWHTPRMAP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(C1=CC=CC=C1)C1=CC(=NC)N(C)C(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)