Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Span 60 (=Sorbitan Monostearate), TCI America™
SDP

Catalog No. S0062500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0062500G 500 g
S006225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0062500G Supplier TCI America Supplier No. S0062500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
Synonym sorbitan stearate, sorbitan stearate inn, span 60 tn, sorbitan monostearate nf
PubChem CID 23725022
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Melting Point 52°C
Color Yellow
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Span 60 (=Sorbitan Monostearate)
EINECSNumber (8)-0063
RTECSNumber WG2933500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.