accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133970-31-7

OONH(S)OHONHOOCl

CAS RN 133970-31-7

IUPAC Name: (2S)-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid|FMOC-LYS(2-CL-Z)-OH
Molecular Weight (g/mol): 537.01
Molecular Formula: C29H29ClN2O6
InChi Key: VUEYAXRHPZGZOL-SANMLTNESA-N
SMILES: OC(=O)[C@H](CCCCNC(=O)OCC1=CC=CC=C1Cl)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)